Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat SIRP alpha/CD172a Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7307G
This item is not returnable.
View return policy
Description
SIRP alpha/CD172a Monoclonal specifically detects SIRP alpha/CD172a in Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
SIRP alpha/CD172a | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFR, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPA, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
140885 | |
Mouse, Rat | |
Purified |
Flow Cytometry | |
772734 | |
Flow Cytometry 5 uL/10^6 cells | |
P97710 | |
SIRPA | |
Mouse myeloma cell line NS0-derived recombinant rat SIRP alpha/CD172a Lys32-Asn373 Accession # P97710 | |
100 Tests | |
Primary | |
Detects rat SIRP alpha/CD172a in direct ELISAs and Western blots. Detects mouse and rat SIRP alpha/CD172a in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction