Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT1 Mouse anti-Human, Mouse, Clone: 10-E-4, MilliporeSigma™

Catalog No. 041557MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-155-7MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-155-7MI Supplier MilliporeSigma Supplier No. 041557
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-SIRT1 Antibody, clone 10E4 (mouse monoclonal antibody) validated in western blot, immunoprecipitation, immunocytochemistry, immunohistochemistry, Chimmunoprecipitation to detect SIRT1 also known as sirtuin (silent mating type information regulation 2 homolog) 1, sirtuin 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRT1
Applications ChIP Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10-E-4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgGκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_036370.2
Gene Symbols SIRT1, SIR2L1, hSIR2, hSIRT1, SIR2alpha
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to human SIRT1.
Purification Method Protein G purfied
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.