Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirt1 Mouse anti-Human, Clone: 3H10.2, MilliporeSigma™

Catalog No. 051243MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-124-3MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-124-3MI Supplier MilliporeSigma Supplier No. 051243
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Sirt1 Antibody, clone 3H10.2 is a Mouse Monoclonal Antibody for detection of Sirt1 also known as Sirtuin 1, Silent mating type Information Regulation-2 and has been tested in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sirt1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3H10.2
Conjugate Unconjugated
Formulation Purified in PBS with 0.05% Sodium Azide
Gene Accession No. NM_012238
Gene Symbols SIRT1, SIR2L1, hSIR2, OTTHUMP00000060745, hSIRT1, SIR2alpha
Host Species Mouse
Immunogen GST fusion protein corresponding to amino acids 508-616 of human Sirt1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.