Learn More
sirtuin (silent mating type information regulation 2 homolog) 1 (S. cerevisiae), Mouse, Clone: 7B7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SIRT1.
Supplier: Abnova Corporation H00023411M01
Description
This gene encodes a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class I of the sirtuin family. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: NRYIFHGAEVYSDSEDDVLSSSSCGSNSDSGTCQSPSLEEPMEDESEIEEFYNGLEDEPDVPERAGGAGFGTDGDDQEAINEAISVKQEVTDMNYPSNKSSpecifications
sirtuin (silent mating type information regulation 2 homolog) 1 (S. cerevisiae) | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC012499 | |
SIRT1 | |
SIRT1 (AAH12499, 456 a.a. ∼ 555 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
7B7 | |
Mouse monoclonal antibody raised against a partial recombinant SIRT1. | |
SIRT1 | |
SIR2L1 | |
Mouse | |
Affinity Purified | |
RUO | |
23411 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.