Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirt1(Sir2) Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™

Catalog No. 07131MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-131-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-131-MI Supplier MilliporeSigma Supplier No. 07131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-Sirt1(Sir2) Antibody (Rabbit Polyclonal Antibody) is published (more than 30 citations) and validated in immunocytochemistry, immunoprecipitation, western blotting to detect Sirt1(Sir2) also known as Silencing information regulator 2-like, sirtuin 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sirt1(Sir2)
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit IgG in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% sodium azide and 30% gylcerol
Gene Symbols SIR2L1; SIR2alpha; hSIR2; hSIRT1
Host Species Rabbit
Immunogen GST-tagged fusion protein corresponding to amino acids 1-131 of mouse Sir2α.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_019812
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.