Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT2 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 09843MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-843-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-843-MI Supplier MilliporeSigma Supplier No. 09843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-SIRT2 Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunoprecipitation to detect SIRT2 also known as SIR2-like protein 2, silent information regulator 2, sirtuin 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRT2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols SIR2; SIR2-like; SIR2L; SIR2L2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the mature protein of human SIRT2.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_036369
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.