Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIVA1, apoptosis-inducing factor (B01P), Mouse anti-Human, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017006 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-006 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-006 Supplier Abnova Corporation Supplier No. H00010572B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SIVA protein.

This gene encodes a protein with an important role in the apoptotic (programmed cell death) pathway induced by the CD27 antigen, a member of the tumor necrosis factor receptor (TFNR) superfamily. The CD27 antigen cytoplasmic tail binds to the N-terminus of this protein. Two alternatively spliced transcript variants encoding distinct proteins have been described. [provided by RefSeq

Sequence: MPKRSCPFADVAPLQLKVRVSQRELSRGVCAERYSQEVFDPSGVASIACSSCVRPVDGKAVCGQCERALCGQCVRTCWGCGSVACTLCGLVDCSDMYEKVLCTSCAMFET

Specifications

Antigen SIVA1, apoptosis-inducing factor
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SIVA protein.
Formulation PBS with no preservative; pH 7.4
Gene SIVA1
Gene Accession No. BC034562
Gene Alias CD27BP/SIVA/Siva-1/Siva-2
Gene Symbols SIVA1
Host Species Mouse
Immunogen SIVA (AAH34562, 1 a.a. ∼ 110 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10572
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.