Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-SIX6, Polyclonal, Invitrogen™
Supplier: Thermo Scientific PA117189
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Hu, Rt, Ms
Immunogen: Synthetic peptide based on the sequence: VLSQGSGRALRAEGDG
Immunocytochemistry, Immunohistochemistry, Western Blotting

Specifications
SIX6 | |
Unconjugated | |
Synthetic peptide based on the sequence: VLSQGSGRALRAEGDG | |
Primary | |
Purified |
Polyclonal | |
Rabbit | |
100 μg | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction