Learn More
Valutek™ Nanotek PE Coated Polypropylene Anti-Skid Boot Covers
Catalog No. p-7162548
Click to view available options
Size:
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Features a durable skid-resistant PVC sole design and offers protection from slips or falls as well as being impervious to liquids. Recommended for use in a cleanroom Class 1-10 (ISO 3-4) critical environments.
  • Clean, low-linting, and latex-free fabric
  • Non-skid PVC sole provides protection against wet and smooth surfaces
  • Coefficient of friction
Specifications
Size Large
Material Polypropylene
Latex Free Yes
Coating Polyethylene
Color White
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type Embossed PVC
Packaging Quantity 200/Cs.
Packaging Type 20/Bag, 10 Bags/Cs.
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 1-10 (ISO 3-4)
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.