Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Valutek™ MacroTek Anti-Skid Polypropylene Shoe Covers
                    Click to view available options
                
                
                    
                            
                                Size:
                            
                        
                        
                            
                                Medium
                            
                        
                            
                                Large
                            
                        
                            
                                X-Large
                            
                        
                    Description
- All Valutek™ shoe covers are tested and manufactured in ISO compliant facilities under individual inspection and strict process control to ensure Valutek™ quality standard and product specifications
 - Recommended for use in a cleanroom Class 10,000+ (ISO 7+) critical environment
 - Commonly used for protecting against dirt, grime, and certain dry particulates in nonhazardous environments
 
Specifications
Specifications
| Size | Medium | 
| Material | Polypropylene | 
| Color | Blue | 
| Disposable | Single-use | 
| Sterility | Non-sterile | 
| Closure Type | Elastic Ankle | 
| Packaging Quantity | 300/Cs. | 
| Packaging Type | 100/Bag, 3 Bags/Cs. | 
| Product Type | Controlled Environments Shoe Covers | 
| Cleanroom Class | Class 10,000 (ISO 7) | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction