Learn More
Src-like-adaptor 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SLA2 protein.
Supplier: Abnova Corporation H00084174D01P
Description
This gene encodes a member of the SLAP family of adapter proteins. The encoded protein may play an important receptor-proximal role in downregulating T and B cell-mediated responses and inhibits antigen receptor-induced calcium mobilization. This protein interacts with Cas-Br-M (murine) ecotropic retroviral transforming sequence c. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: MGSLPSRRKSLPSPSLSSSVQGQGPVTMEAERSKATAVALGSFPAGGPAELSLRLGEPLTIVSEDGDWWTVLSEVSGREYNIPSVHVAKVSHGWLYEGLSREKAEELLLLPGNPGGAFLIRESQTRRGSYSLSVRLSRPASWDRIRHYRIHCLDNGWLYISPRLTFPSLQALVDHYSELADDICCLLKEPCVLQRAGPLPGKDIPLPVTVQRTPLNWKELDSSLLFSEAATGEESLLSEGLRESLSFYISLNDEAVSLDDASpecifications
| Src-like-adaptor 2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human SLA2 protein. | |
| SLA2 | |
| C20orf156/FLJ21992/MGC49845/SLAP-2/SLAP2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_032214 | |
| SLA2 | |
| SLA2 (NP_115590.1, 1 a.a. ∼ 261 a.a) full-length human protein. | |
| 100 μg | |
| Membrane Receptors | |
| 84174 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.