Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008108 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-108 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-108 Supplier Abnova Corporation Supplier No. H00064849B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SLC13A3 protein.

Mammalian sodium-dicarboxylate cotransporters transport succinate and other Krebs cycle intermediates. They fall into 2 categories based on their substrate affinity: low affinity and high affinity. Both the low- and high-affinity transporters play an important role in the handling of citrate by the kidneys. The protein encoded by this gene represents the high-affinity form. Alternatively spliced transcript variants encoding different isoforms have been found for this gene, although the full-length nature of some of them have not been characterized yet. [provided by RefSeq

Sequence: MAALAAAAKKVWSARRLLVLLFTPLALLPVVFALPPKEGRCLFVILLMAVYWCTEALPLSVTALLPIVLFPFMGILPSNKVCPQYFLDTNFLFLSGLIMASAIEEWNLHRRIALKILMLVGVQPARLILGMMVTTSFLSMWLSNTASTAMMLPIANAILKSLFGQKEVRKDPSQESEENTAAVRRNGLHTVPTEMQFLASTEAKDHPGETEVPLDLPADSRKEDEYRRNIWKGFLISIPYSASIGGTATLTGTAPNLILLGQLKSFFPQCDVVNFGSWFIFAFPLMLLFLLAGWLWISFLYGGLSFRGWRKNKSEIRTNAEDRARAVIREEYQNLGPIKFAEQAVFILFCMFAILLFTRDPKFIPGWASLFNPGFLSDAVTGVAIVTILFFFPSQRPSLKWWFDFKAPNTETEPLLTWKKAQETVPWNIILLLGGGFAMAKGCEESGLSVWIGGQLHPLENVPPALAVLLITVVIAFFTEFASNTATIIIFLPVLAELAIRLRVHPLYLMIPGTVGCSFAFMLPVSTPPNSIAFASGHLLVKDMVRTGLLMNLMGVLLLSLAMNTWAQTIFQLGTFPDWADMYSVNVTALPPTLANDTFRTL

Specifications

Antigen solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SLC13A3 protein.
Formulation No additive
Gene SLC13A3
Gene Accession No. NM_022829.4
Gene Alias NADC3/SDCT2
Gene Symbols SLC13A3
Host Species Mouse
Immunogen SLC13A3 (NP_073740.2, 1 a.a. ∼ 602 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 64849
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.