Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
solute carrier family 14 (urea transporter), member 1 (Kidd blood group), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SLC14A1 protein.
Supplier: Abnova Corporation H00006563B01P
Description
The protein encoded by this gene is a membrane transporter that mediates urea transport in erythrocytes. This gene forms the basis for the Kidd blood group system. [provided by RefSeq
Sequence: MEDSPTMVRVDSPTMVRGENQVSPCQGRRCFPKALGYVTGDMKELANQLKDKPVVLQFIDWILRGISQVVFVNNPVSGILILVGLLVQNPWWALTGWLGTVVSTLMALLLSQDRSLIASGLYGYNATLVGVLMAVFSDKGDYFWWLLLPVCAMSMTCPIFSSALNSVLSKWDLPVFTLPFNMALSMYLSATGHYNPFFPAKLVIPITTAPNISWSDLSALELLKSIPVGVGQIYGCDNPWTGGIFLGAILLSSPLMCLHAAIGSLLGIAAGLSLSAPFENIYFGLWGFNSSLACIAMGGMFMALTWQTHLLALGCALFTAYLGVGMANFMAEVGLPACTWPFCLATLLFLIMTTKNSNIYKMPLSKVTYPEENRIFYLQAKKRMVESPLSpecifications
solute carrier family 14 (urea transporter), member 1 (Kidd blood group) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human SLC14A1 protein. | |
SLC14A1 | |
FLJ33745/FLJ41687/HUT11/HsT1341/JK/RACH1/UT-B1/UT1/UTE | |
Mouse | |
Affinity Purified | |
RUO | |
6563 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC050539 | |
SLC14A1 | |
SLC14A1 (AAH50539, 1 a.a. ∼ 389 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction