Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC18A1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89003080 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-080 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-080 Supplier Abnova Corporation Supplier No. H00006570A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant SLC18A1.

The vesicular monoamine transporter acts to accumulate cytosolic monoamines into vesicles, using the proton gradient maintained across the vesicular membrane. Its proper function is essential to the correct activity of the monoaminergic systems that have been implicated in several human neuropsychiatric disorders. The transporter is a site of action of important drugs, including reserpine and tetrabenazine (Peter et al., 1993 [PubMed 7905859]). See also SLC18A2 (MIM 193001).[supplied by OMIM

Sequence: MLRPILDAPQRLLKEGRASRQLVLVVVFVALLLDNMLFTVVVPIVPTFLYDMEFKEVNSSLHLGHAGSSPHALASPAFSTIFSFFNNNTVAVEESVPSGIAWMNDTASTIPPPATEAISAHKNNCLQGTGFLEEETTRVGVLFASKAVMQLLVNPFVGPLTNRIGYHIPMFAGFVIMFLSTVMFAFSGTYTLLFVARTLQGIGSSFSSVAGLGMLASVYTDDHERGRAMGTALGGLALGLLVGAPFGSVMYEFVGKSAPFLILAFLALLDGALQLCILQPSKVSPESAKGTPLFMLLKDPYILVAAGLAFLPASVSYLIGTNLFGVLANKMGRWLCSLIGMLVVGTSLLCVPLAHNIFGLIGPNAGLGLAIGMVDSSMMPIMGHLVDLRHTSVYGSVYAIADVAFCMGFAIGPSTGGAIVKAIGFPWLMVITGVINIVYAPLCYYLRSPPAKEEKLAILSQDCPMETRMYATQKPTKEFPLGEDSDEEPDHEE

Specifications

Antigen SLC18A1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant SLC18A1.
Formulation 50% glycerol
Gene SLC18A1
Gene Accession No. BC006317
Gene Alias CGAT/VAT1/VMAT1
Gene Symbols SLC18A1
Host Species Mouse
Immunogen SLC18A1 (AAH06317.1, 1 a.a. ∼ 493 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6570
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.