Learn More
solute carrier family 18 (vesicular monoamine), member 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLC18A2.
Supplier: Abnova Corporation H00006571A01
Description
The vesicular monoamine transporter acts to accumulate cytosolic monoamines into synaptic vesicles, using the proton gradient maintained across the synaptic vesicular membrane. Its proper function is essential to the correct activity of the monoaminergic systems that have been implicated in several human neuropsychiatric disorders. The transporter is a site of action of important drugs, including reserpine and tetrabenazine (Peter et al., 1993 [PubMed 7905859]). See also SLC18A1 (MIM 193002).[supplied by OMIM
Sequence: HEKNATEIQTARPVHTASISDSFQSIFSYYDNSTMVTGNATRDLTLHQTATQHMVTNASAVPSDCPSEDKDLLNENVQVGLLFASKATVQLITNPFIGLSpecifications
solute carrier family 18 (vesicular monoamine), member 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SLC18A2. | |
SLC18A2 | |
MGC120477/MGC120478/MGC26538/SVAT/SVMT/VAT2/VMAT2 | |
Mouse | |
50 μL | |
Yes | |
6571 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_003054 | |
SLC18A2 | |
SLC18A2 (NP_003045, 53 a.a. ∼ 151 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.