Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

solute carrier family 1 (glial high affinity glutamate transporter), member 2, Mouse, Clone: 4G8, Abnova™

Catalog No. 89014189 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-189 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-189 Supplier Abnova Corporation Supplier No. H00006506M10
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SLC1A2.

This gene encodes a member of a family of solute transporter proteins. The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system. Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors. Mutations in and decreased expression of this protein are associated with amyotrophic lateral sclerosis. Alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. [provided by RefSeq

Sequence: DEVSSLDAFLDLIRNLFPENLVQACFQQIQTVTKKVLVAPPPDEEANATSAVVSLLNETVTEVPEETKMVIKKGLEFKDG

Specifications

Antigen solute carrier family 1 (glial high affinity glutamate transporter), member 2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SLC1A2.
Formulation PBS with no preservative; pH 7.4
Gene SLC1A2
Gene Accession No. NM_004171
Gene Alias EAAT2/GLT-1
Gene Symbols SLC1A2
Host Species Mouse
Immunogen SLC1A2 (NP_004162, 160 a.a. ∼ 239 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6506
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.