Learn More
SLC25A5, Mouse, Clone: 1D7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SLC25A5.
Supplier: Abnova Corporation H00000292M01
Description
This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein forms a homodimer embedded in the inner mitochondria membrane. Suppressed expression of this gene has been shown to induce apoptosis and inhibit tumor growth. The human genome contains several nontranscribed pseudogenes of this gene
Sequence: MTDAAVSFAKDFLAGGVAAAISKTAVAPIERVKLLLQVQHASKQITADKQYKGIIDCVVRIPKEQGVLSFWRGNLANVIRYFPTQALNFAFKDKYKQIFLGGVDKRTQFWRYFAGNLASGGAAGATSLCFVYPLDFARTRLAADVGKAGAEREFRGLGDCLVKIYKSDGIKGLYQGFNVSVQGIIIYRAAYFGIYDTAKGMLPDPKNTHIVISWMIAQTVTAVAGLTSYPFDTVRRRMMMQSGRKGTDIMYTGTLDCWRKIARDEGGKAFFKGAWSNVLRGMGGAFVLVLYDEIKKYTSpecifications
SLC25A5 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC056160 | |
SLC25A5 | |
SLC25A5 (AAH56160, 1 a.a. ∼ 298 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
1D7 | |
Mouse monoclonal antibody raised against a full-length recombinant SLC25A5. | |
SLC25A5 | |
2F1/AAC2/ANT2/T2/T3 | |
Mouse | |
Affinity chromatography | |
RUO | |
292 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.