Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC26A2, Mouse, Clone: 3F6, Abnova™

Catalog No. 89021361 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-361 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-361 Supplier Abnova Corporation Supplier No. H00001836M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SLC26A2.

The diastrophic dysplasia sulfate transporter is a transmembrane glycoprotein implicated in the pathogenesis of several human chondrodysplasias. It apparently is critical in cartilage for sulfation of proteoglycans and matrix organization. [provided by RefSeq

Sequence: MSSESKEQHNVSPRDSAEGNDSYPSGIHLELQRESSTDFKQFETNDQCRPYHRILIERQEKSDTNFKEFVIKKLQKNCQCSPAKAKNMILGFLPVLQWLPKYDLKKNI

Specifications

Antigen SLC26A2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SLC26A2.
Formulation PBS with no preservative; pH 7.4
Gene SLC26A2
Gene Accession No. NM_000112
Gene Alias D5S1708/DTD/DTDST/EDM4/MST153/MSTP157
Gene Symbols SLC26A2
Host Species Mouse
Immunogen SLC26A2 (NP_000103.1, 1 a.a. ∼ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1836
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.