Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

solute carrier family 26 (sulfate transporter), member 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016770 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-770 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-770 Supplier Abnova Corporation Supplier No. H00001836B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SLC26A2 protein.

The diastrophic dysplasia sulfate transporter is a transmembrane glycoprotein implicated in the pathogenesis of several human chondrodysplasias. It apparently is critical in cartilage for sulfation of proteoglycans and matrix organization. [provided by RefSeq

Sequence: MSSESKEQHNVSPRDSAEGNDSYPSGIHLELQRESSTDFKQFETNDQCRPYHRILIERQEKSDTNFKEFVIKKLQKNCQCSPAKAKNMILGFLPVLQWLPKYDLKKNILGDVMSGLIVGILLVPQSIAYSLLAGQEPVYGLYTSFFASIIYFLLGTSRHISVGIFGVLCLMIGETVDRELQKAGYDNAHSAPSLGMVSNGSTLLNHTSDRICDKSCYAIMVGSTVTFIAGVYQVAMGFFQVGFVSVYLSDALLSGFVTGASFTILTSQAKYLLGLNLPRTNGVGSLITTWIHVFRNIHKTNLCDLITSLLCLLVLLPTKELNEHFKSKLKAPIPIELVVVVAATLASHFGKLHENYNSSIAGHIPTGFMPPKVPEWNLIPSVAVDAIAISIIGFAITVSLSEMFAKKHGYTVKANQEMYAIGFCNIIPSFFHCFTTSAALAKTLVKESTGCHTQLSGVVTALVLLLVLLVIAPLFYSLQKSVLGVITIVNLRGALRKFRDLPKMWSISRMDTVIWFVTMLSSALLSTEIGLLVGVCFSIFCVILRTQKPKSSLLGLVEESEVFESVSAYKNLQTKPGIKIFRFVAPLYYINKECFKSALYKQTVNPILIKVAWKKAAKRKIKEKVVTLGGIQDEMSVQLSHDPLELHTIVIDCSAIQFLDTAGIHTLKEVRRDYEAIGIQVLLAQCNPTVRDSLTNGEYCKKEEENLLFYSVYEAMAFAEVSKNQKGVCVPNGLSLSSD

Specifications

Antigen solute carrier family 26 (sulfate transporter), member 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SLC26A2 protein.
Formulation PBS with no preservative; pH 7.4
Gene SLC26A2
Gene Accession No. BC059390.1
Gene Alias D5S1708/DTD/DTDST/EDM4/MST153/MSTP157
Gene Symbols SLC26A2
Host Species Mouse
Immunogen SLC26A2 (AAH59390.1, 1 a.a. ∼ 739 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1836
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.