Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

solute carrier family 30 (zinc transporter), member 9, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89006985 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-985 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-985 Supplier Abnova Corporation Supplier No. H00010463B01

Mouse polyclonal antibody raised against a full-length human SLC30A9 protein.

Sequence: MLPGLAAAAAHRCSWSSLCRLRLRCRAAACNPSDRQEWQNLVTFGSFSNVVPCSHPYIGTLSQVKLYSTNVQKEGQGSQTLRVEKVPSFETAEGIGAELKAPLKQEPLQVRVKAVLKKREYGSKYTQNNFITGVRAINEFCLKSSDLEQLRKIRRRSPHEDTESFTVYLRSDVEAKSLEVWGSPEALAREKKLRKEAEIEYRERLFRNQKILREYRDFLGNTKPRSRTASVFFKGPGKVVMVAICINGLNCFFKFLAWIYTGSASMFSEAIHSLSDTCNQGLLALGISKSVQTPDPSHPYGFSNMRYISSLISGVGIFMMGAGLSWYHGVMGLLHPQPIESLLWAYCILAGSLVSEGATLLVAVNELRRNARAKGMSFYKYVMESRDPSTNVILLEDTAAVLGVIIAATCMGLTSITGNPLYDSLGSLGVGTLLGMVSAFLIYTNTEALLGRSIQPEQVQRLTELLENDPSVRAIHDVKATDLGLGKVRFKAEVDFDGRVVTRSYLEKQDFDQMLQEIQEVKTPEELETFMLKHGENIIDTLGAEVDRLEKELKKRNPEVRHVDLEIL

Specifications

Antigen solute carrier family 30 (zinc transporter), member 9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SLC30A9 protein.
Formulation No additive
Gene SLC30A9
Gene Accession No. BC007732.2
Gene Alias C4orf1/GAC63/HUEL/ZNT9
Gene Symbols SLC30A9
Host Species Mouse
Immunogen SLC30A9 (AAH07732.1, 1 a.a. ∼ 568 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10463
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.