Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC35A1, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89210723 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-210-723 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-210-723 Supplier Abnova Corporation Supplier No. H00010559K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human SLC35A1 peptide using ARM Technology.

The SLC35A1 gene encodes a CMP-sialic acid transporter located within the membrane of the Golgi apparatus. The transporter moves nucleotide sugars across the membrane for use in glycosylation reactions that take place within the Golgi department (Eckhardt et al., 1996 [PubMed 8755516]). For background information on the SLC35 family of nucleotide-sugar transporters, see SLC35A3 (MIM 605632).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen SLC35A1
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description solute carrier family 35 (CMP-sialic acid transporter), member A1
Formulation In 1X PBS, pH 7.4
Gene SLC35A1
Gene Accession No. SLC35A1
Gene Alias CMPST, CST, hCST, inactive
Gene Symbols SLC35A1
Host Species Rabbit
Immunogen A synthetic peptide of human SLC35A1
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10559
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.