Learn More
SLC3A2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human SLC3A2 peptide using ARM Technology.
Supplier: Abnova Corporation H00006520K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene is a member of the solute carrier family and encodes a cell surface, transmembrane protein with an alpha amylase domain. The protein exists as the heavy chain of a heterodimer, covalently bound through di-sulfide bonds to one of several possible light chains. It associates with integrins and mediates integrin-dependent signaling related to normal cell growth and tumorigenesis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
 - Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
 
Specifications
| SLC3A2 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| SLC3A2 | |
| SLC3A2 | |
| A synthetic peptide of human SLC3A2 | |
| RUO | |
| 6520 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG | 
| ELISA, Western Blot | |
| 1mg | |
| solute carrier family 3 (activators of dibasic and neutral amino acid transport), member 2 | |
| SLC3A2 | |
| 4F2, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid | 
Your input is important to us. Please complete this form to provide feedback related to the content on this product.