Learn More
SLC40A1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human SLC40A1 peptide using ARM Technology.
Supplier: Abnova Corporation H00030061K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is a cell membrane protein that may be involved in iron export from duodenal epithelial cells. Defects in this gene are a cause of hemochromatosis type 4 (HFE4). [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
SLC40A1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
SLC40A1 | |
SLC40A1 | |
A synthetic peptide of human SLC40A1 | |
RUO | |
30061 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
solute carrier family 40 (iron-regulated transporter), member 1 | |
SLC40A1 | |
FPN1, HFE4, IREG1, MST079, MSTP079, MTP1, SLC11A3 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.