Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLFN11 Mouse anti-Human, Clone: 4G9, MilliporeSigma™
GSA_VA

Catalog No. MABF248MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF248MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF248MI Supplier MilliporeSigma Supplier No. MABF248
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-SLFN11 Antibody, clone 4G9 is an antibody against SLFN11 for use in western blotting and immunoprecipitation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLFN11
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4G9
Concentration 0.25mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_001098057
Gene Symbols SLFN11
Host Species Mouse
Immunogen Recombinant protein corresponding to human SLFN11.
Purification Method Purified by ion-exchange chromatography
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.