Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slick potassium channel Mouse anti-Mouse, Rat, Clone: N11/33, MilliporeSigma™

Catalog No. MABN510MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN510MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN510MI Supplier MilliporeSigma Supplier No. MABN510

Mouse Monoclonal Antibody

Anti-Slick potassium channel Antibody, clone N11/33 is a highly specific mouse monoclonal antibody, that targets Slo2.1 and has been tested in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Slick potassium channel
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone N11/33
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001074496
Gene Symbols Kcnt2
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse Slick potassium channel.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.