Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLITRK1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen SLITRK1
Concentration 0.65 mg/mL
Dilution Western Blot 1:2500, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:10-1:500
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP149541SS
SDP
View Documents
Novus Biologicals
NBP149541SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP149541
SDP
View Documents
Novus Biologicals
NBP149541
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

SLITRK1 Polyclonal specifically detects SLITRK1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

SLITRK1
Western Blot 1:2500, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:10-1:500
Polyclonal
Rabbit
Neuroscience
PBS and 30% Glycerol with 0.05% Sodium Azide
FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM
SLITRK1
IgG
Affinity Purified
Human and mouse. Immunogen sequence has 84% homology to rat.
0.65 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse
Q96PX8, Q810C1
114798
A genomic peptide made to an internal region of the human Slitrk1 protein (within residues 250-400). [Swiss-Prot Q96PX8]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.