Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLP-76 (pY128) Mouse anti-Human, Unlabeled, Clone: J141-668.36.58, BD

Catalog No. BDB558367 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558367 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558367 Supplier BD Biosciences Supplier No. 558367
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLP-76 (SH2 domain-containing Leukocyte Protein of 76kDa) is a tyrosine phosphoprotein that is involved in the T cell receptor (TCR) -mediated intracellular signaling pathway. It may be involved in the signaling pathways of other peripheral blood leukocytes; thymic/splenic cells; and in human T, B, and monocytic cell lines. SLP-76 consists of several motifs that signify its importance in protein-protein interactions involved in intracellular signaling pathways, such as the SH2 domain in the C-terminus, the three amino-terminus 17-amino acid repeats with conserved tyrosine and acidic residues (DYE(S/P)P), and a proline rich region. SLP-76 has been shown to associate with Gads, Grb2, PLCg1, SLAP-130, and Vav, all of which are part of the signaling cascade in T lymphocytes. An early event in the T cell activation pathway is the phosphorylation, by the Syk-family kinase ZAP-70, of SLP-76 at the three conserved tyrosine motifs, which then mediate interactions with downstream effectors. The phosphorylated tyrosine 128 (Y128) brings the Rho family guanine nucleotide exchange factor Vav1 and the Nck adapter protein, which binds to p21-activated kinase (PAK1) and Wiskott-Aldrich syndrome protein (WASP), into the activation complex. Vav1, PAK1, and WASP may mediate TCR-stimulated actin cytoskeletal rearrangement. The J141-688.36.58 monoclonal antibody recognizes the phosphorylated Y128 of activated SLP-76.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLP-76 (pY128)
Applications Western Blot
Classification Monoclonal
Clone J141-668.36.58
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human SLP-76
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.