Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad1 (pS463/pS465) Rat, Unlabeled, Clone: N6-1233, BD

Catalog No. BDB562508 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562508 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562508 Supplier BD Biosciences Supplier No. 562508

Rat Monoclonal Antibody

The N6-1233 monoclonal antibody specifically binds to the Smad1 protein phosphorylated at the Ser463/465 sites and the Smad8 protein phosphorylated at the Ser465/467 sites. Smad1 and Smad8 are ∽60 kDa proteins and are members of the Smad superfamily. The Smad family members are divided into three subfamilies: receptor regulated Smads or R-Smads, including Smads1, 2, 3, 5, and 8; common partner Smad, or Co-Smad, including Smad4; and inhibitory Smads, or I-Smad, including Smads 6 and 7. Activation of Transforming Growth Factor-beta (TGF-beta) superfamily serine/threonine kinase receptors, such as TGF-beta and Bone MorphogenicProtein (BMP) receptors, leads to the phosphorylation of R-Smads at several sites. It has been shown that Ser463 and Ser465 of Smad1 are phosphorylated by BMP receptors. In B cells and pre-B cells, BMP-6 has been shown to induce Smad1/5/8 phosphorylation and inhibit cell proliferation. Phosphorylated R-Smads form complexes with Co-Smad and translocate into the nucleus to regulate transcription affecting a wide range of critical processes including cell-fate determination, proliferation, morphogenesis, differentiation and apoptosis. The inhibitory Smads inhibit this pathway through two potential mechanisms: either by preventing R-Smads from binding to their corresponding receptors and/or by competing with Smad4, the Co-Smad, from binding to R-Smads. This antibody may crossreact with Smad5 pS463/pS465 based on sequence homology.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Smad1 (pS463/pS465)
Applications Western Blot
Classification Monoclonal
Clone N6-1233
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Smad1,Smad8/Smad9; MADH1, MADH9
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Phosphorylated Human Smad1 aa 456-465 Peptide
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.