Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smad2/3 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07408MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-408-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-408-MI Supplier MilliporeSigma Supplier No. 07408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Smad2/3 Antibody is a Rabbit Polyclonal Antibody for detection of Smad2/3 also known as MAD-mothers against decapentaplegic-homolog 2 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Smad2/3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol.
Gene Symbols SMAD2; MGC22139; MGC34440; MADR2; hMAD-2; JV18; hSMAD2; MADH2; JV18-1
Host Species Rabbit
Immunogen Fusion protein corresponding to amino acids 186-273 of human Smad2.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001003652
Target Species Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.