Learn More
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SMARCA4.
Supplier: Abnova Corporation H00006597A01
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins and is similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. In addition, this protein can bind BRCA1, as well as regulate the expression of the tumorigenic protein CD44. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: LSPNPPNLTKKMKKIVDAVIKYKDSSSGRQLSEVFIQLPSRKELPEYYELIRKPVDFKKIKERIRNHKYRSLNDLEKDVMLLCQNAQTFNLEGSLIYEDSSpecifications
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SMARCA4. | |
SMARCA4 | |
BAF190/BRG1/FLJ39786/SNF2/SNF2-BETA/SNF2L4/SNF2LB/SWI2/hSNF2b | |
Mouse | |
50 μL | |
Transcription Regulation | |
6597 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
Unconjugated | |
50% glycerol | |
NM_003072 | |
SMARCA4 | |
SMARCA4 (NP_003063, 1451 a.a. ∼ 1550 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.