Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-SMC3, Polyclonal,
Supplier: MilliporeSigma™ AB3914
Description

Specifications
SMC3 | |
Polyclonal | |
Affinity purified immunoglobulin. Liquid in Tris-citrate/phosphate, pH 7-8 with 0.1% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Xenopus species |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
SMC3; bamacan; BMH; HCAP; BAM; CDLS3; CSPG6; SMC3L1; Bamacan; hCAP | |
Synthetic peptide from human SMC-3. | |
50 μg | |
NM_005445.3 | |
2-8°C in undiluted aliquots for up to 6 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction