Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SMN2 Ab-1, MaxPab™ Polyclonal, Abnova™

Catalog No. 89002914 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-914 Supplier Abnova Corporation Supplier No. H00006607D01
Add to Cart
Add to Cart

Rabbit Polyclonal

Host Species: Rabbit
Cross Reactivity: Human
Immunogen: SMN2 (NP_075013.1, 1 a.a. ∽282 a.a) full-length human protein.

Detection Antibody, Immunoprecipitation

Specifications

Antigen SMN2
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SMN2 protein.
Formulation No additive
Gene SMN2
Gene Accession No. NM_022875
Gene Alias BCD541/C-BCD541/FLJ76644/MGC20996/MGC5208/SMNC
Gene Symbols SMN2
Host Species Rabbit
Immunogen SMN2 (NP_075013.1, 1 a.a. ∼ 282 a.a) full-length human protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6607
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.