Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SMN2 Ab-2, Polyclonal, Abnova™

Catalog No. 89003082 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-082 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-082 Supplier Abnova Corporation Supplier No. H00006607A01
Add to Cart
Add to Cart

Mouse Polyclonal

Host Species: Mouse
Cross Reactivity: Human
Immunogen: SMN2 (NP_059107, 191 a.a. ∽ 295 a.a) partial recombinant protein with GST tag

ELISA, Western Blotting

Specifications

Antigen SMN2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SMN2.
Formulation 50% glycerol
Gene SMN2
Gene Accession No. NM_017411
Gene Alias BCD541/C-BCD541/FLJ76644/MGC20996/MGC5208/SMNC
Gene Symbols SMN2
Host Species Mouse
Immunogen SMN2 (NP_059107, 191 a.a. ∼ 294 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6607
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.