Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SMN2, Clone: 1A3-2B9, Abnova™

Catalog No. 89123293 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-293 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-293 Supplier Abnova Corporation Supplier No. H00006607M02
Add to Cart
Add to Cart

Mouse Monoclonal

Host Species: Mouse
Clone: 1A3-2B9
Isotype: IgG2a, κ
Cross Reactivity: Human
Immunogen: SMN2 (AAH00908, 1 a.a. ∽282 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa

ELISA, Sandwich ELISA, Western Blotting

Specifications

Antigen SMN2
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1A3-2B9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant SMN2.
Formulation PBS with no preservative; pH 7.4
Gene SMN2
Gene Accession No. BC000908
Gene Alias BCD541/C-BCD541/FLJ76644/MGC20996/MGC5208/SMNC
Gene Symbols SMN2
Host Species Mouse
Immunogen SMN2 (AAH00908, 1 a.a. ∼ 282 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6607
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.