Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase), Mouse, Clone: 2C9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SMPD2.
Supplier: Abnova Corporation H00006610M01
Description
Sequence: MKPNFSLRLRIFNLNCWGIPYLSKHRADRMRRLGDFLNQESFDLALLEEVWSEQDFQYLRQKLSPTYPAAHHFRSGIIGSGLCVFSKHPIQELTQHIYTLNGYPYMIHHGDWFSGKAVGLLVLHLSGMVLNAYVTHLHAEYNRQKDIYLAHRVAQAWELAQFIHHTSKKADVVLLCGDLNMHPEDLGCCLLKEWTGLHDAYLETRDFKGSEEGNTMVPKNCYVSQQELKPFPFGVRIDYVLYKAVSGFYISCKSFETTTGFDPHSGTPLSDHEALMATLFVRHSPPQQNPSSTHGPAERSPLMCVLKEAWTELGLGMAQARWWATFASYVIGLGLLLLALLCVLAAGGGAGEAAILLWTPSVGLVLWAGAFYLFHVQEVNGLYRAQAELQHVLGRAREAQDLGPEPQPALLLGQQEGDRTKEQSpecifications
sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase) | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC000038 | |
SMPD2 | |
SMPD2 (AAH00038.1, 1 a.a. ∼ 423 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
50 μg | |
Endocrine/Metabolism | |
6610 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG3 κ |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
2C9 | |
Mouse monoclonal antibody raised against a full length recombinant SMPD2. | |
SMPD2 | |
NSMASE/NSMASE1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction