Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

single-strand-selective monofunctional uracil-DNA glycosylase 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004567 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-567 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-567 Supplier Abnova Corporation Supplier No. H00023583A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SMUG1.

SMUG1 is a glycosylase that removes uracil from single- and double-stranded DNA in nuclear chromatin, thus contributing to base excision repair.[supplied by OMIM

Sequence: PQAFLLGSIHEPAGALMEPQPCPGSLAESFLEEELRLNAELSQLQFSEPVGIIYNPVEYAWEPHRNYVTRYCQGPKE

Specifications

Antigen single-strand-selective monofunctional uracil-DNA glycosylase 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SMUG1.
Formulation 50% glycerol
Gene SMUG1
Gene Accession No. NM_014311
Gene Alias FDG/HMUDG/MGC104370/UNG3
Gene Symbols SMUG1
Host Species Mouse
Immunogen SMUG1 (NP_055126, 2 a.a. ∼ 78 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 23583
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.