Learn More
SMAD specific E3 ubiquitin protein ligase 1, Mouse, Clone: 1D7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SMURF1.
Supplier: Abnova Corporation H00057154M01
Description
This gene encodes a ubiquitin ligase that is specific for receptor-regulated SMAD proteins in the bone morphogenetic protein (BMP) pathway. A similar protein in Xenopus is involved in embryonic pattern formation. Alternative splicing results in multiple transcript variants encoding different isoforms. An additional transcript variant has been identified, but its full length sequence has not been determined. [provided by RefSeq
Sequence: DSGPGRPLSCFMEEPAPYTDSTGAAAGGGNCRFVESPSQDQRLQAQRLRNPDVRGSLQTPQNRPHGHQSPELPEGYEQRTTVQGQVYFLHTQTGVSTWHDPRIPSpecifications
SMAD specific E3 ubiquitin protein ligase 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_020429 | |
SMURF1 | |
SMURF1 (NP_065162, 165 a.a. ∼ 268 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
1D7 | |
Mouse monoclonal antibody raised against a partial recombinant SMURF1. | |
SMURF1 | |
KIAA1625 | |
Mouse | |
Affinity Purified | |
RUO | |
57154 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.