Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAD specific E3 ubiquitin protein ligase 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018554 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-554 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-554 Supplier Abnova Corporation Supplier No. H00057154D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human SMURF1 protein.

This gene encodes a ubiquitin ligase that is specific for receptor-regulated SMAD proteins in the bone morphogenetic protein (BMP) pathway. A similar protein in Xenopus is involved in embryonic pattern formation. Alternative splicing results in multiple transcript variants encoding different isoforms. An additional transcript variant has been identified, but its full length sequence has not been determined. [provided by RefSeq

Sequence: MSNPGTRRNGSSIKIRLTVLCAKNLAKKDFFRLPDPFAKIVVDGSGQCHSTDTVKNTLDPKWNQHYDLYVGKTDSITISVWNHKKIHKKQGAGFLGCVRLLSNAISRLKDTGYQRLDLCKLNPSDTDAVRGQIVVSLQTRDRIGTGGSVVDCRGLLENEGTVYEDSGPGRPLSCFMEEPAPYTDSTGAAAGGGNCRFVESPSQDQRLQAQRLRNPDVRGSLQTPQNRPHGHQSPELPEGYEQRTTVQGQVYFLHTQTGVSTWHDPRIPRDLNSVNCDELGPLPPGWEVRSTVSGRIYFVDHNNRTTQFTDPRLHHIMNHQCQLKEPSQPLPLPSEGSLEDEELPAQRYERDLVQKLKVLRHELSLQQPQAGHCRIEVSREEIFEESYRQIMKMRPKDLKKRLMVKFRGEEGLDYGGVAREWLYLLCHEMLNPYYGLFQYSTDNIYMLQINPDSSINPDHLSYFHFVGRIMGLAVFHGHYINGGFTVPFYKQLLGKPIQLSDLESVDPELHKSLVWILENDITPVLDHTFCVEHNAFGRILQHELKPNGRNVPVTEENKKEYVRLYVNWRFMRGIEAQFLALQKGFNELIPQHLLKPFDQKELELIIGGLDKIDLNDWKSNTRLKHCVADSNIVRWFWQAVETFDEERRARLLQFVTGSTRVPLQGFKALQGSTGAAGPRLFTIHLIDANTDNLPKAHTCFNRIDIPPYESYEKLYEKLLTAVEETCGFAVE

Specifications

Antigen SMAD specific E3 ubiquitin protein ligase 1
Applications In situ PLA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human SMURF1 protein.
Formulation PBS with no preservative; pH 7.4
Gene SMURF1
Gene Accession No. NM_181349
Gene Alias KIAA1625
Gene Symbols SMURF1
Host Species Rabbit
Immunogen SMURF1 (NP_851994.1, 1 a.a. ∼ 731 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57154
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.