Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAI1 Mouse anti-Rat, Mouse, Human, Clone: 10H4.1, MilliporeSigma™

Catalog No. MABE167MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE167MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE167MI Supplier MilliporeSigma Supplier No. MABE167
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-SNAI1 Antibody, clone 10H4.1 is a highly specific mouse monoclonal antibody, that targets Zinc Finger Protein and has been tested in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAI1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 10H4.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_005976
Gene Symbols SNAI1, SNAH
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of Human SNAIL1.
Purification Method Purified by ion-exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.