Learn More
SNAI1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human SNAI1 peptide using ARM Technology.
Supplier: Abnova Corporation H00006615K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The Drosophila embryonic protein snail is a zinc finger transcriptional repressor which downregulates the expression of ectodermal genes within the mesoderm. The nuclear protein encoded by this gene is structurally similar to the Drosophila snail protein, and is also thought to be critical for mesoderm formation in the developing embryo. At least two variants of a similar processed pseudogene have been found on chromosome 2. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
SNAI1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
SNAI1 | |
SNAI1 | |
A synthetic peptide of human SNAI1 | |
RUO | |
6615 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
snail homolog 1 (Drosophila) | |
SNAI1 | |
SLUGH2, SNA, SNAH, dJ710H13.1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.