Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAP-25 Mouse, Unlabeled, Clone: 20, BD

Catalog No. BDB610366 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610366 50 μg
BDB610367 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610366 Supplier BD Biosciences Supplier No. 610366
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Release of neurotransmitters from neurons is regulated by exocytosis of synaptic vesicles. This exocytosis is mediated by a complex consisting of membrane components of both the synaptic vesicle and the synaptic plasma membrane. The fusion complex consists of the soluble NSF (N-ethyl-maleimide-sensitive factor) and SNAPs (soluble NSF attachment proteins), along with the receptor proteins (known as SNAREs) synaptobrevin, synaptotagmin, syntaxin, and SNAP-25 (synaptosomal-associated protein of 25kDa- the name is coincidental to the previously mentioned “SNAP” terminology). SNAP-25 and syntaxin are plasmalemmal proteins (designated as t-SNAREs) while synaptobrevin and synaptotagmin are vesicular proteins (designated as v-SNAREs). These four proteins are thought to constitute an initial SNARE docking complex for regulated exocytosis. SNAP-25 lacks a transmembrane domain, but is linked to the membrane by palmitoylated cysteine residues in the central region of the molecule.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP-25
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Description Synaptosomal Associated Protein of 25 kD
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.