Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAPC2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535172
Description
SNAPC2 is a subunit of the snRNA-activating protein complex which is associated with the TATA box-binding protein. The encoded protein is necessary for RNA polymerase II and III dependent small-nuclear RNA gene transcription. Alternative splicing results in multiple transcript variants.
Specifications
SNAPC2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q13487 | |
SNAPC2 | |
KLH conjugated synthetic peptide between 278-307 amino acids from the C-terminal region of human SNAPC2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SNAPC2 | |
SNAPC2; SNAP45; snRNA-activating protein complex subunit 2; Proximal sequence element-binding transcription factor subunit delta; Small nuclear RNA-activating complex polypeptide 2; snRNA-activating protein complex 45 kDa subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6618 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction