Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
small nuclear RNA activating complex, polypeptide 5, 19kDa, Mouse, Clone: 5E2, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SNAPC5.
Supplier: Abnova Corporation H00010302M03
Description
Sequence: MLSRLQELRKEEETLLRLKAALHDQLNRLKMLVHVDNEASINQTTLELSTKSHVTEEEEEEEEEESDSSpecifications
small nuclear RNA activating complex, polypeptide 5, 19kDa | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC014315 | |
SNAPC5 | |
SNAPC5 (AAH14315, 1 a.a. ∼ 68 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
5E2 | |
Mouse monoclonal antibody raised against a full length recombinant SNAPC5. | |
SNAPC5 | |
SNAP19 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
10302 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction