Learn More
Description
Host Species: Mouse
Cross Reactivity: Human, Rat
Immunogen: SNCAIP (AAH94759.1, 1 a.a. ∽603 a.a) full-length human protein.
Detection Antibody, Western Blotting
Specifications
Specifications
| Antigen | SNCAIP |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | SNCAIP (AAH94759.1, 1 a.a. ∽603 a.a) full-length human protein. |
| Quantity | 50 μL |
| Primary or Secondary | Primary |
| Target Species | Human, Rat |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.