Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNCB, Mouse, Clone: 4F11, Abnova™

Catalog No. 89123296 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-296 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-296 Supplier Abnova Corporation Supplier No. H00006620M16
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SNCB.

Sequence: MDVFMKGLSMAKEGVVAAAEKTKQGVTEAAEKTKEGVLYVGSKTREGVVQGVASVAEKTKEQASH

Specifications

Antigen SNCB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4F11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SNCB.
Formulation PBS with no preservative; pH 7.4
Gene SNCB
Gene Accession No. NM_001001502
Gene Symbols SNCB
Host Species Mouse
Immunogen SNCB (NP_001001502.1, 1 a.a. ∼ 65 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6620
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.