Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNF2Β/BRG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07478MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-478-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-478-MI Supplier MilliporeSigma Supplier No. 07478
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-SNF2β/BRG1 Antibody (rabbit polyclonal antibody) validated in immunoprecipitation, western blotting to detect SNF2β/BRG1 also known as Probable global transcription activator SNF2L4, ATP-dependent helicase SMARCA4, SNF2-beta, BRG-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNF2β/BRG1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal in buffer containing with 0.05% sodium azide with 30% glycerol.
Gene Symbols SMARCA4; BRG1; SNF2B; SNF2L4
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the Near N-Terminus of human SNF2β.
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003072
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.