Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNF2L Rat anti-Human, Clone: SNF 2C4, MilliporeSigma™

Catalog No. MABE366MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE366MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE366MI Supplier MilliporeSigma Supplier No. MABE366
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect the SNF2L protein using this Anti-SNF2L, clone SNF 2C4 validated for use in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNF2L
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone SNF 2C4
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_003060
Gene Symbols SMARCA1, SNF2L, SNF2L1
Host Species Rat
Immunogen GST-tagged recombinant protein corresponding to human SNF2L.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.