Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNF related kinase, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007766 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-766 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-766 Supplier Abnova Corporation Supplier No. H00054861B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human SNRK protein.

SNRK is a member of the sucrose nonfermenting (SNF)-related kinase family of serine/threonine kinases (Kertesz et al., 2002 [PubMed 12234663]).[supplied by OMIM

Sequence: MAGFKRGYDGKIAGLYDLDKTLGRGHFAVVKLARHVFTGEKVAVKVIDKTKLDTLATGHLFQEVRCMKLVQHPNIVRLYEVIDTQTKLYLILELGDGGDMFDYIMKHEEGLNEDLAKKYFAQIVHAISYCHKLHVVHRDLKPENVVFFEKQGLVKLTDFGFSNKFQPGKKLTTSCGSLAYSAPEILLGDEYDAPAVDIWSLGVILFMLVCGQPPFQEANDSETLTMIMDCKYTVPSHVSKECKDLITRMLQRDPKRRAYLEEIENHPWLQGVDPSPATKYNIPLVSYKNLSEEEHNSIIQRMVLGDIADRDAIVEALETNRYNHITATYFLLAERILREKQEKEIQTRSASPSNIKAQFRQSWPTKIDVPQDLEDDLTATPLSHATVPQSPARAADSVLNGHRSKGLCDSAKKDDLPELAGPALSTVPPASLKPTASGRKCLFRVEEDEEEDEEDKKPMSLSTQVVLRRKPSVTNRLTSRKSAPVVNQIFEEGESDDEFDMDENLPPKLSRLKMNIASPGTVHKRYHRRKSQGRGSSCSSSETSDDDSESRRRLDKDSGFTYSWHRRDSSEGPPGSEGDGGGQSKPSNASGGVDKASPSENNAGGGSPSSGSGGNPTNTSGTTRRCAGPSNSMQLASRSAGELVESLKLMSLCLGSQLHGSTKYIIDPQNGLSFSSVKVQEKSTWKMCISSTGNAGQVPAVGGIKFFSDHMADTTTELERIRSKNLKNNVLQLPLCEKTISVNIQRNPKEGLLCASSPASCCHVI

Specifications

Antigen SNF related kinase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human SNRK protein.
Formulation No additive
Gene SNRK
Gene Accession No. BC071567.1
Gene Alias DKFZp779A1866/FLJ20224/HSNFRK/KIAA0096
Gene Symbols SNRK
Host Species Mouse
Immunogen SNRK (AAH71567.1, 1 a.a. ∼ 765 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 54861
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.