Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRNP35, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89206960 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-206-960 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-206-960 Supplier Abnova Corporation Supplier No. H00011066K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human SNRNP35 peptide using ARM Technology.

The protein encoded by this gene is a homolog of U1-snRNP binding protein. The N-terminal half contains a RNA recognition motif and the C-terminal is rich in Arg/Asp and Arg/Glu dipeptides; a characteristic of a variety of splicing factors. This protein is a component of the U11/U12 small nuclear ribonucleoproteins (snRNP) that form part of the U12-type spliceosome. This gene is differentially expressed in a variety of human tissues. Alternative splicing results in multiple transcript variants encoding distinct proteins

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen SNRNP35
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description small nuclear ribonucleoprotein 35kDa (U11/U12)
Formulation In 1X PBS, pH 7.4
Gene SNRNP35
Gene Accession No. SNRNP35
Gene Alias HM-1, MGC138160, U1SNRNPBP
Gene Symbols SNRNP35
Host Species Rabbit
Immunogen A synthetic peptide of human SNRNP35
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11066
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.