Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNRNP40 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525196
Description
This antibody is predicted to react with bovine and mouse based on sequence homology.
SNRNP40 encodes a component of the U5 small nuclear ribonucleoprotein (snRNP) particle. The U5 snRNP is part of the spliceosome, a multiprotein complex that catalyzes the removal of introns from pre-messenger RNAs.
Specifications
SNRNP40 | |
Polyclonal | |
SNRNP40 | |
40K, HPRP8BP, PRP8BP, PRPF8BP, RP11-490K7.3, SPF38, WDR57, 38 kDa-splicing factor, Prp8-binding protein, U5 small nuclear ribonucleoprotein 40 kDa protein, U5 snRNP 40 kDa protein, U5 snRNP-specific 40 kDa protein (hPrp8-binding), U5-40K, U5-40kD protein, WD repeat domain 57 (U5 snRNP specific), WD repeat-containing prot | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
66585, 9410 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q96DI7, Q6PE01 | |
SNRNP40 | |
KLH conjugated synthetic peptide between 122-149 amino acids from the central region of human SNRNP40 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction